  • Search Results
  • Identification of a positively charged platform in Staphylococcus aureus HtsA that is essential for ferric staphyloferrin A transport.
Identification of a positively charged platform in Staphylococcus aureus HtsA that is essential for ferric staphyloferrin A transport.
Biochemistry (2014-07-23)
John D Cooper, Mélissa Hannauer, Cristina L Marolda, Lee-Ann K Briere, David E Heinrichs
ABSTRACT
In response to iron starvation, Staphylococcus aureus secretes both staphyloferrin A and staphyloferrin B, which are high-affinity iron-chelating molecules. The structures of both HtsA and SirA, the ferric-staphyloferrin A [Fe(III)-SA] and ferric-staphyloferrin B [Fe(III)-SB] receptors, respectively, have recently been determined. The structure of HtsA identifies a novel form of ligand entrapment composed of many positively charged residues. Through ionic interactions, the binding pocket appears highly adapted for the binding of the highly anionic siderophore SA. However, biological validation of the importance of the nine SA-interacting residues (six arginines, one tyrosine, one histidine, and one lysine) has not been previously performed. Here, we mutated each of the Fe(III)-SA-interacting residues in HtsA and found that substitutions R104A, R126A, H209A, R306A, and R306K resulted in a reduction of binding affinity of HtsA for Fe(III)-SA. While mutation of almost all proposed ligand-interacting residues decreased the ability of S. aureus cells to transport (55)Fe(III)-SA, S. aureus expressing HtsA R104A, R126A, R306A, and R306K showed the greatest transport defects and were incapable of growth in iron-restricted growth media in a SA-dependent manner. These three residues cluster together and, relative to other residues in the binding pocket, move very little between the apo and closed holo structures. Their essentiality for receptor function, together with structural information, suggests that they form a positively charged platform that is required for initial contact with the terminal carboxyl groups of the two citrates in the Fe(III)-SA complex. This is a likely mechanism by which HtsA discerns iron-bound SA from iron-free SA.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Mandelic acid, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Supelco
DL-Mandelic acid, analytical standard
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)