  • Search Results
  • A galloylated dimeric proanthocyanidin from grape seed exhibits dentin biomodification potential.
A galloylated dimeric proanthocyanidin from grape seed exhibits dentin biomodification potential.
Fitoterapia (2014-12-30)
Rasika S Phansalkar, Joo-Won Nam, Shao-Nong Chen, James B McAlpine, José G Napolitano, Ariene Leme, Cristina M P Vidal, Thaiane Aguiar, Ana K Bedran-Russo, Guido F Pauli
ABSTRACT
Grape seeds are a rich source of polyphenols, especially proanthocyanidins (PACs), and are also known for the presence of galloylated oligomeric PACs (OPACs). The present study focuses on the phytochemical methodology for grape seed (O)PACs and their potential role as dentin biomodifiers to be used in restorative and reparative dentistry. A new method using centrifugal partition chromatography (CPC) was developed for the preparative separation of the grape seed (O)PACs. Orthogonal phytochemical profiling of the resulting CPC fractions was performed using C18 and diol HPLC, normal phase HPTLC, and IT-TOF MS analysis. A galloylated procyanidin dimer (1) was isolated from a CPC fraction in order to evaluate its potential to enhance dentin bio-mechanical properties. Moreover, it helped to evaluate the impact of the galloyl moiety on the observed bioactivity. Structure elucidation was performed using ESI-MS, 1D and 2D NMR analyses. For the first time, (1)H iterative full spin analysis (HiFSA) was performed on this type of molecule, enabling a detailed proton chemical shift and coupling constant assignment. The CPC fractions as well as 1 showed promising results in the dentin stiffness bioassay and indicate that they may be used as dental intervention biomaterial.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Gallic acid, certified reference material, TraceCERT®
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Sigma-Aldrich
Methanol, anhydrous, 99.8%