SPAD aptasensor for the detection of circulating protein biomarkers.
Biosensors & bioelectronics (2015-01-31)
Laura Pasquardini, Lucio Pancheri, Cristina Potrich, Alessandro Ferri, Claudio Piemonte, Lorenzo Lunelli, Lucia Napione, Valentina Comunanza, Maria Alvaro, Lia Vanzetti, Federico Bussolino, Cecilia Pederzolli
ABSTRACT
The need for decentralized clinical tests together with the concept of time and cost saving are pushing the development of portable, miniaturized, compact biosensors with diagnostic and prognostic purpose. Here, we propose an innovative detection system based on a Single Photon Avalanche Diode (SPAD) with high sensitivity and low noise, crucial features for an efficient chemiluminescence biosensor. The SPAD detector, having 60 µm diameter, has a Photon Detection Efficiency higher than 55% at 425 nm and a Dark Count Rate lower than 100 Hz at room temperature. Our design allows a good optical coupling efficiency between sample and detector. A specific biofunctional surface was implemented taking advantage of aptamers, short DNA sequences having high selectivity and affinity toward their targets. We successfully detected physiological levels of Vascular Endothelial Growth Factor (VEGF), a circulating protein biomarker highly correlated with cancer. The SPAD aptasensor showed a Limit of Detection (LoD) in the pM range, stability (up to 42 days) and re-usability (up to seven cycles). This compact biosensor is therefore a promising step toward the actual use of portable microdevices in diagnostics.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)