  • Search Results
  • Thioredoxin 1 upregulates FOXO1 transcriptional activity in drug resistance in ovarian cancer cells.
Thioredoxin 1 upregulates FOXO1 transcriptional activity in drug resistance in ovarian cancer cells.
Biochimica et biophysica acta (2014-12-09)
Jianlin Wang, Hao Yang, Wenjie Li, Huibi Xu, Xiangliang Yang, Lu Gan
ABSTRACT
Drug resistance is the major cause of failure of cancer chemotherapy in ovarian cancer. However, the molecular mechanisms on the regulation of drug resistance are not fully understood. Here we showed that Trx1 and FOXO1 were involved in paclitaxel (PTX)-induced drug resistance in ovarian cancer A2780 cells. PTX induced reactive oxygen species (ROS) and resulted in Trx1 and FOXO1 nuclear translocation. We further found that Trx1 bound to FOXO1 and enhanced FOXO1 transcriptional activity; however Trx1 C69S mutant which is barely detected in the nucleus downregulated Trx1-FOXO1 interaction and Trx1-induced FOXO1 transcriptional activation. Silencing of FOXO1 abrogated Trx1-induced drug resistance. Trx1 increased FOXO1-induced drug resistance, while Trx1 C69S mutant completely abolished the regulation of FOXO1-mediated drug resistance by Trx1. These findings provided a novel mechanism on Trx1/FOXO1 signaling in drug resistance in ovarian cancer cells.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
2′,7′-Dichlorofluorescin diacetate, ≥97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent