  • Search Results
  • Combining De Ley-Doudoroff and methylerythritol phosphate pathways for enhanced isoprene biosynthesis from D-galactose.
Combining De Ley-Doudoroff and methylerythritol phosphate pathways for enhanced isoprene biosynthesis from D-galactose.
Bioprocess and biosystems engineering (2014-06-15)
Kristine Rose M Ramos, Kris Niño G Valdehuesa, Huaiwei Liu, Grace M Nisola, Won-Keun Lee, Wook-Jin Chung
ABSTRACT
An engineered Escherichia coli strain was developed for enhanced isoprene production using D-galactose as substrate. Isoprene is a valuable compound that can be biosynthetically produced from pyruvate and glyceraldehyde-3-phosphate (G3P) through the methylerythritol phosphate pathway (MEP). The Leloir and De Ley-Doudoroff (DD) pathways are known existing routes in E. coli that can supply the MEP precursors from D-galactose. The DD pathway was selected as it is capable of supplying equimolar amounts of pyruvate and G3P simultaneously. To exclusively direct D-galactose toward the DD pathway, an E. coli ΔgalK strain with blocked Leloir pathway was used as the host. To obtain a fully functional DD pathway, a dehydrogenase encoding gene (gld) was recruited from Pseudomonas syringae to catalyze D-galactose conversion to D-galactonate. Overexpressions of endogenous genes known as MEP bottlenecks, and a heterologous gene, were conducted to enhance and enable isoprene production, respectively. Growth test confirmed a functional DD pathway concomitant with equimolar generation of pyruvate and G3P, in contrast to the wild-type strain where G3P was limiting. Finally, the engineered strain with combined DD-MEP pathway exhibited the highest isoprene production. This suggests that the equimolar pyruvate and G3P pools resulted in a more efficient carbon flux toward isoprene production. This strategy provides a new platform for developing improved isoprenoid producing strains through the combined DD-MEP pathway.
Sigma-Aldrich
Thiamine pyrophosphate, ≥95%
Supelco
Thiamine pyrophosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Supelco
Isoprene, analytical standard
SAFC
Isoprene, ≥99%
Sigma-Aldrich
Isoprene, 99%, contains <1000 ppm p-tert-butylcatechol as inhibitor
Sigma-Aldrich
D-(+)-Galactose, BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), ≥99%
Sigma-Aldrich
D-(+)-Galactose, ≥99%
Sigma-Aldrich
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98%
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard
SAFC
Galactose, plant-derived
USP
Galactose, United States Pharmacopeia (USP) Reference Standard