  • Search Results
  • Fiber supplementation lowers plasma p-cresol in chronic kidney disease patients.
Fiber supplementation lowers plasma p-cresol in chronic kidney disease patients.
Journal of renal nutrition : the official journal of the Council on Renal Nutrition of the National Kidney Foundation (2014-12-03)
Younis A Salmean, Mark S Segal, Sergiu P Palii, Wendy J Dahl
ABSTRACT
To determine the effects of supplemental fiber on plasma p-cresol, stool frequency, and quality of life (QoL) in chronic kidney disease (CKD) patients. In a 12-week single-blind study, participants were provided with control muffins and supplements (5.5 g sucrose/day) for 2 weeks, muffins containing 10 g/day pea hull fiber and control supplements for 4 weeks, and muffins with 10 g/day pea hull fiber and 15 g/day inulin as a supplement for 6 weeks. Individuals with CKD (n = 13; 6 males, 7 females; aged 65 ± 3 years; estimated glomerular filtration rate <50 mL/minute/1.73(2)) completed the study. Plasma p-cresol was determined by gas chromatography-mass spectrometry, stool frequency by 5-day journals, and QoL by the KDQOL-36™. Plasma p-cresol decreased from 7.25 ± 1.74 mg/L during week 1 to 5.82 ± 1.72 mg/L during week 12 (P < .05), and in participants with high compliance (>70% inulin intake), from 6.71 ± 1.98 mg/L to 4.22 ± 1.16 mg/L (P < .05). Total fiber intake increased from 16.6 ± 1.7 g/day during control to 26.5 ± 2.4 g/day (P < .0001) with the added pea hull and to 34.5 ± 2.2 g/day with pea hull and inulin (P < .0001). Stool frequency increased from 1.4 ± 0.2 stools/day during control to 1.9 ± 0.3 stools/day during both fiber periods (P < .05). No change in overall QoL was observed. Supplementing the diet of CKD patients with fiber may be a dietary therapy to reduce p-cresol and improve stool frequency.
Sigma-Aldrich
p-Cresol, ≥99%, FG
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
p-Cresol, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
p-Cresol, 99%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)