  • Search Results
  • An improved method for the characterization of supersaturation and precipitation of poorly soluble drugs using pulsatile microdialysis (PMD).
An improved method for the characterization of supersaturation and precipitation of poorly soluble drugs using pulsatile microdialysis (PMD).
International journal of pharmaceutics (2014-04-09)
Kosha B Shah, Piyush G Patel, Akm Khairuzzaman, Robert A Bellantone
ABSTRACT
In current pharmaceutical drug discovery, most candidates are poorly soluble in water, which can result in poor bioavailability. To overcome this problem, formulations that create supersaturation of the drug are a well-studied alternative. Characterizing the dissolution from these systems is challenging because conventional methods, such as sampling with a syringe then filtering with a 0.2-0.45 μm filter before an HPLC assay, can overestimate the concentration of dissolved drug by allowing nuclei or small precipitated particles to pass, which then dissolve in the HPLC mobile phase. Nuclei and small particles can also cause overestimation of the dissolved concentration when using optical methods. Such overestimations can lead to failure of in vivo prediction of drug bioavailability from supersaturated systems. This paper reports a novel method to determine the free dissolved drug concentration in a dissolution medium using pulsatile microdialysis (PMD). Ibuprofen was used as a model drug for determining precipitation and supersaturation. Supersaturation was induced chemically by changing pH, and also by dissolution/release from an in-house formulation. The adaptation of a previously developed PMD model is summarized, and experimental results comparing dissolved concentrations determined using PMD and direct sampling by syringe and filtering are presented.
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)