  • Search Results
  • Cloning and characterization of a phytoene dehydrogenase gene from marine yeast Rhodosporidium diobovatum.
Cloning and characterization of a phytoene dehydrogenase gene from marine yeast Rhodosporidium diobovatum.
Antonie van Leeuwenhoek (2015-01-30)
Wenjing Guo, Yuxuan Liu, Xin Yan, Mingyi Liu, Hui Tang, Zheng Liu, Liping Zhang
ABSTRACT
Carotenoids are a naturally occurring and widely distributed group of pigments that provide protection against photooxidation and inactivate free radicals due to their highly conjugated double-bond systems. Phytoene dehydrogenation is the first rate-limiting step in the carotenoid biosynthetic pathway. Phytoene dehydrogenase is the key enzyme in the transformation of carotenoid from colorless to colored; therefore it is the first target of gene manipulation. The present study describes the identification and functional characterization of a carontenoid synthesis gene from Rhodosporidium diobovatum, designated as crtI, which catalyzes the dehydrogenation of phytoene. We obtained a full-length cDNA clone of crtI, encoding phytoene dehydrogenase (EC Number: 1.3.99.31), from R. diobovatum ATCC 2527 by rapid amplification of cDNA ends. Complementation mapping of the crtI gene in Escherichia coli allowed us to localize the regions responsible for phytoene dehydrogenase function within the protein. Enzyme activity of the expressed protein in E. coli was verified using high performance liquid chromatography analysis. We were able to determine the nucleotide sequence of crtI from R. diobovatum. The publicly available sequence will be useful in future studies on phytoene dehydrogenase.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%