  • Search Results
  • Quantification and visualization of glutathione S-transferase omega 1 in cells using inductively coupled plasma mass spectrometry (ICP-MS) and fluorescence microscopy.
Quantification and visualization of glutathione S-transferase omega 1 in cells using inductively coupled plasma mass spectrometry (ICP-MS) and fluorescence microscopy.
Analytical and bioanalytical chemistry (2014-11-21)
Yong Liang, Xin Jiang, Nannan Tang, Limin Yang, Haifeng Chen, Qiuquan Wang
ABSTRACT
We report a novel activity-based and Cu-free click chemistry (CC) mediated methodology for glutathione S-transferase omega 1 (GSTO1) quantification using species-unspecific isotope dilution inductively coupled plasma mass spectrometry (SUID ICP-MS), in which dibenzylcyclooctyne-modified 2-chloroacetamide (DBCO-ChAcA) was designed and synthesized, meanwhile, as a navigator towards GSTO1 for subsequent N3-DOTA-Eu-tagging via Cu-free CC. Using (153)Eu-SUID ICP-MS coupled with size exclusion chromatography (SEC), the LOD (3σ) of GSTO1 reached 6.9 fmol with an RSD of 2.4% at the 0.1 μM level (n = 5) considering the recovery of GSTO1 on the SEC was 96.5 ± 2.4%. The GSTO1 contents in the cells of human hepatocellular carcinoma C7721 and breast carcinoma MCF-7 as well as normal hepatic C7701 without or with cis-platin administration were quantified to be from 1.2 μg/10,000 cells (n = 3, RSD = 4.5%) corresponding to 1.2 × 10(-2) ng per cell to 4.76 μg/10,000 cells (n = 3, RSD = 2.9%) corresponding to 4.76 × 10(-2) ng per cell. For a comparative study, DBCO-ChAcA-fluor 488-based fluorescence microscopy could not alone visualize GSTO1 in the cells but could together with those from the small SH-containing molecules such as GSH and that from extra N3-fluor 488 in the cells. This activity-based CC-mediated tagging/labeling strategy provided an opportunity for ICP-MS-based targeted protein quantification, and is very much expected to find its applications in biological mechanism study and the subsequent drug design.
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), One-time steel-plastic (SP) drum
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Supelco
trans-Sinapic acid, analytical standard
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), Customized plastic drum
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)