  • Search Results
  • Simultaneous determination of remimazolam and its carboxylic acid metabolite in human plasma using ultra-performance liquid chromatography-tandem mass spectrometry.
Simultaneous determination of remimazolam and its carboxylic acid metabolite in human plasma using ultra-performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-09)
Ying Zhou, Hongyun Wang, Ji Jiang, Pei Hu
ABSTRACT
A robust and validated method based on ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS) has been developed for the simultaneous determination of remimazolam, which is a new chemical entity, and its major carboxylic acid metabolite (M1) in human plasma. Plasma samples were pre-purified by protein precipitation procedure and analyzed using an isocratic chromatographic separation over an Acquity UPLC CSH C18 column. The mobile phase consisted of acetonitrile-water containing 10mM ammonium formate and 0.1% formic acid at a flow rate of 0.4mLmin(-1). Positive electrospray ionization was employed as the ionization source in the multiple reaction monitoring (MRM) mode. The analysis time was about 1.5min. The method was fully validated over the concentration range of 0.5-1000ngmL(-1) for both analytes. The lower limit of quantification (LLOQ) was 0.5ngmL(-1). Inter- and intra-batch precision was less than 8.4% and the accuracy was within 88.8-107.0%. The mean extraction recoveries obtained from three concentrations of QC plasma samples were 96.8%, 98.7% and 98.6% for remimazolam, 98.7%, 99.8% and 101.5% for M1, respectively. Selectivity, matrix effect and stability were also validated. The method was applied to the pharmacokinetic study of remimazolam in Chinese healthy subjects.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%