  • Search Results
  • Synthesis, characterization and mechanistic-insight into the anti-proliferative potential of PLGA-gemcitabine conjugate.
Synthesis, characterization and mechanistic-insight into the anti-proliferative potential of PLGA-gemcitabine conjugate.
International journal of pharmaceutics (2014-05-09)
Vaibhav Khare, Smit Kour, Noor Alam, Ravindra Dharr Dubey, Ankit Saneja, Mytre Koul, Ajai Prakash Gupta, Deepika Singh, Shashank K Singh, Ajit K Saxena, Prem N Gupta
ABSTRACT
Gemcitabine, a nucleoside analogue, is used in the treatment of various solid tumors, however, its efficacy is limited by rapid metabolism by cytidine deaminase and fast kidney excretion. In this study, a polymeric conjugate of gemcitabine was prepared by covalent coupling with poly(lactic-co-glycolic) acid (PLGA), in order to improve anticancer efficacy of the drug. The prepared conjugate was characterized by various analytical techniques including FTIR, NMR and mass spectroscopic analysis. The stability study indicated that the polymeric conjugate was more stable in plasma as compared to native gemcitabine. Further, in vitro cytotoxicity determined in a panel of cell lines including pancreatic cancer (MIAPaCa-2), breast cancer (MCF-7) and colon cancer (HCT-116), indicated that the cytotoxic activity of gemcitabine was retained following conjugation with polymeric carrier. In the nucleoside transportation inhibition assay, it was found that the prepared conjugate was not dependent on nucleoside transporter for entering into the cells and this, in turn, reflecting potential implication of this conjugate in the therapy of transporter- deficient resistance cancer. Further, the cell cycle analysis showed that the sub-G1 (G0) apoptotic population was 46.6% and 60.6% for gemcitabine and PLGA gemcitabine conjugate, respectively. The conjugate produced remarkable decrease in mitochondrial membrane potential, a marker of apoptosis. In addition, there was a marked increase in PARP cleavage and P-H2AX expression with PLGA gemcitabine conjugate as compared to native gemcitabine indicating improved apoptotic activity. The findings demonstrated the potential of PLGA gemcitabine conjugate to improve clinical outcome of gemcitabine based chemotherapy of cancer.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dichloromethane, analytical standard
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
N,N′-Dicyclohexylurea, 98%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Supelco
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Millipore
Dichloromethane, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)