  • Search Results
  • Characterization and hepatoprotective effect of polysaccharides from Ziziphus jujuba Mill. var. spinosa (Bunge) Hu ex H. F. Chou sarcocarp.
Characterization and hepatoprotective effect of polysaccharides from Ziziphus jujuba Mill. var. spinosa (Bunge) Hu ex H. F. Chou sarcocarp.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-09-27)
Yuan Yue, Shuangchan Wu, Huifang Zhang, Xuyan Zhang, Yunhui Niu, Xiaoqin Cao, Fuwei Huang, Hong Ding
ABSTRACT
The polysaccharides from Ziziphus jujuba Mill. var. spinosa (Bunge) Hu ex H. F. Chou sarcocarp (PWJS) is evaluated for the chemical composition, antioxidant activity and hepatoprotective effect. The characteristics of PWJS were determined by FT-IR spectral and HPLC analysis. The antioxidant activity was investigated using in vitro systems. An in vivo study of PWJS against CCl4 induced liver injury was also conducted. HPLC analysis showed that PWJS is an acidic heteropolysaccharide, rich in glucose (38.59%), arabinose (23.16%), galacturonic acid (17.64%) and galactose (10.44%). PWJS displayed strong antioxidant activity in vitro. In the in vivo study, PWJS treatment lowered the serum levels of ALT and AST in CCl4-intoxicated mice. Additionally, levels of antioxidant enzymes (SOD and CAT) and GSH were elevated in liver damage mice by PWJS intervention, while content of MDA was lessened. Meanwhile, PWJS reverses the suppression of Nrf2 nuclear translocation, and increases the protein expression of HO-1, GSTα and NQO1 in liver damage mice. Hematoxylin-eosin staining showed that the condition of liver damage was mitigated. This study demonstrates that PWJS reverses hepatotoxicity in CCl4-intoxicated mice through the mechanisms of antioxidant activity, as well as an augmentation of the Nrf2 pathway in liver tissue.
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-(+)-Arabinose, ≥99%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
D-(+)-Galactose, ≥99%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
D-(+)-Galactose, ≥98%
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Ribose, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material