  • Search Results
  • Glycerin esterification of scum derived free fatty acids for biodiesel production.
Glycerin esterification of scum derived free fatty acids for biodiesel production.
Bioresource technology (2015-10-20)
Erik Anderson, Min Addy, Qinglong Xie, Huan Ma, Yuhuan Liu, Yanling Cheng, Nonso Onuma, Paul Chen, Roger Ruan
ABSTRACT
Scum is an oily waste stream of the wastewater treatment process that can be used to produce biodiesel. Combining acid hydrolysis and solvent extraction, a free fatty acid and acyl-glycerol rich product was produced. Free fatty acids (FFAs) present were converted to acyl-glycols via a high temperature (238°C) glycerin esterification process known as glycerolysis. The inorganic catalysts zinc aluminum oxide and sodium sulfate were tested during glycerolysis to compare the reaction kinetics of converting FFA to acyl-glycerols. It was concluded that the zinc-based catalyst increased the reaction rate significantly, from a "k" value of 2.57 (uncatalyzed) to 5.63, completing the reaction in 60min, half the time it took the uncatalyzed reaction (120min). Sodium sulfate's presence however slowed the reaction, resulting in a "k" value of 1.45, completing the reaction in 180min. Use of the external catalyst Zn-Al2O3 showed the greatest catalytic potential, but also assumes additional costs.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Sodium methoxide solution, 25 wt. % in methanol
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)