  • Search Results
  • Enhancing the in vitro anti-cancer efficacy of artesunate by loading into poly-D,L-lactide-co-glycolide (PLGA) nanoparticles.
Enhancing the in vitro anti-cancer efficacy of artesunate by loading into poly-D,L-lactide-co-glycolide (PLGA) nanoparticles.
Archives of pharmacal research (2014-06-28)
Hanh Thuy Nguyen, Tuan Hiep Tran, Jong Oh Kim, Chul Soon Yong, Chien Ngoc Nguyen
ABSTRACT
Artesunate (ART)-a well-known anti-malarial agent is also known to have potential anti-proliferative activities but its instability, poor aqueous solubility, and lack of relevant studies have limited its application as an effective anti-cancer drug. To overcome these problems, ART was loaded in poly (lactic-co-glycolic) acid (PLGA) nanoparticles using oil/water emulsion evaporation method. PLGA nanoparticles with small particle size and high entrapment efficiency were obtained. The PLGA nanoparticles were optimized by evaluating the effects of several formulation parameters on physicochemical properties of nanoparticles. The in vitro cytotoxicity of blank PLGA, free ART, and ART-PLGA on 3 human cancer cell lines viz. A549, SCC-7, and MCF-7 was conducted using MTT assay. The particles showed nanometric size (~170 nm), large entrapment efficiency (up to 83.4%), and excellent stability (evaluated for 1 month) after lyophilization with 5% mannitol. ART was dispersed inside particle core allowing a sustained release up to 48 h. The in vitro cytotoxicity results demonstrated strong activity of ART against cancer cell lines. The ART-PLGA formulation significantly reduced cell viability than the free ART. The formulation of ART loaded PLGA nanoparticles supported a potential application of ART as an anticancer agent.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture