  • Search Results
  • Nutritional and health-promoting properties of bean paste fortified with onion skin in the light of phenolic-food matrix interactions.
Nutritional and health-promoting properties of bean paste fortified with onion skin in the light of phenolic-food matrix interactions.
Food & function (2015-08-26)
Ł Sęczyk, M Swieca, U Gawlik-Dziki
ABSTRACT
The study examined the effect of fortification of bean paste with onion skin phenolics. The antioxidant potential and in vitro digestibility of nutrients in the light of phenolic-food matrix interactions were studied. Bean paste was supplemented with onion skin extracts in the range of 5-50 mg of phenolics per 100 g of bean (C, O5-O50). Fortification increased the phenolic level and antioxidant activity of bean paste; however, the experimental values, in most cases, were significantly lower than those predicted. The digestive tract acted as an effective extractor of phenolics from the functional products but, surprisingly, the amounts of introduced phenolics had no significant effect on its determined levels. Additionally, after in vitro intestinal digestion a significant increase in the ability to quench free radicals was only observed in samples enriched with the highest amounts of phenolics (O50). After gastric and intestinal stages of digestion no significant effect of fortification on the albumin digestibility was observed. Contrarily, digestibility of globulins had decreased from 70% (C) up to 55% (O50) after gastric digestion and from 93% (C) up to 80% (O50) after intestinal digestion. These results were confirmed by SDS-PAGE analysis, which indicates the presence of indigestible complexes after supplementation with phenolics. Moreover, a significant decrease in starch digestibility from 43% (C) to 21% (O50) was found. In conclusion, fortification improves the health-promoting properties of bean paste; however, the interactions of phenolics with the food matrix play an important role in the creation of the nutraceutical and nutritional quality of products.
Sigma-Aldrich
5-(Dimethylamino)-1-naphthalenesulfonamide, 99%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)