  • Search Results
  • Nanostructured Electron-Selective Interlayer for Efficient Inverted Organic Solar Cells.
Nanostructured Electron-Selective Interlayer for Efficient Inverted Organic Solar Cells.
ACS applied materials & interfaces (2015-08-05)
Jiyun Song, Jaehoon Lim, Donggu Lee, M Thambidurai, Jun Young Kim, Myeongjin Park, Hyung-Jun Song, Seonghoon Lee, Kookheon Char, Changhee Lee
ABSTRACT
We report a unique nanostructured electron-selective interlayer comprising of In-doped ZnO (ZnO:In) and vertically aligned CdSe tetrapods (TPs) for inverted polymer:fullerene bulkheterojunction (BHJ) solar cells. With dimension-controlled CdSe TPs, the direct inorganic electron transport pathway is provided, resulting in the improvement of the short circuit current and fill factor of devices. We demonstrate that the enhancement is attributed to the roles of CdSe TPs that reduce the recombination losses between the active layer and buffer layer, improve the hole-blocking as well as electron-transporting properties, and simultaneously improve charge collection characteristics. As a result, the power conversion efficiency of PTB7:PC70BM based solar cell with nanostructured CdSe TPs increases to 7.55%. We expect this approach can be extended to a general platform for improving charge extraction in organic solar cells.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexylamine, 99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
SAFC
Hexadecyltrimethylammonium bromide
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Vetec, reagent grade, 96%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
1,8-Diiodooctane, 98%, contains copper as stabilizer
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
SAFC
Ethanolamine