  • Search Results
  • Exposure of the population of Catalonia (Spain) to musk fragrances through seafood consumption: Risk assessment.
Exposure of the population of Catalonia (Spain) to musk fragrances through seafood consumption: Risk assessment.
Environmental research (2015-04-29)
Laura Trabalón, German Cano-Sancho, Eva Pocurull, Martí Nadal, José L Domingo, Francesc Borrull
ABSTRACT
The occurrence of ten synthetic musks in samples of 10 widely consumed fish and shellfish species from Tarragona (Catalonia, Spain) was determined. The most used nitro and polycyclic musks, as well as a well-known transformation product in tissues, were analyzed. Furthermore, the human health risks derived from the musk exposure through seafood consumption were characterized. None of the nitro musks were detected in any of the analyzed samples. In contrast, most of the polycyclic musks were found, being galaxolide (HHCB) and tonalide (AHTN) present in all the samples. HHCB was the greatest contributor, with maximum levels in sardine and mackerel (367 and 304 ng g(-1) (d.w.) (dry weight), respectively). The highest exposure to individual musks was estimated for HHCB and HHCB-Lactone, with average values of 19.7 and 6.8 ng kg(-1)bw day(-1), respectively, in adults. A notably lower mean exposure was calculated for AHTN, cashmeran (DPMI) and traseolide (ATII), being ranged between 1.1 and 3.7 ng kg(-1)bw day(-1). The current concentrations of musks in fish and shellfish should not mean human health risks for the adult population living in Tarragona. However, a continuous monitoring would be desirable to assure that the exposure does not follow increasing temporal trends.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), Customized plastic drum
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), One-time steel-plastic (SP) drum
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ethyl acetate