  • Search Results
  • Gastroprotective Activities of Sennoside A and Sennoside B via the Up-Regulation of Prostaglandin E2 and the Inhibition of H(+)/K(+)-ATPase.
Gastroprotective Activities of Sennoside A and Sennoside B via the Up-Regulation of Prostaglandin E2 and the Inhibition of H(+)/K(+)-ATPase.
Biomolecules & therapeutics (2015-09-04)
In Young Hwang, Choon Sik Jeong
ABSTRACT
Sennoside A (erythro) and sennoside B (threo) are dianthrone glycosides and diastereomers. We investigated their abilities to prevent the gastric lesions associated with diseases, such as, gastritis and gastric ulcer. To elucidate their gastroprotective effects, the inhibitions of HCl•EtOH-induced gastritis and indomethacin-induced gastric ulcers were assessed in rats. It was observed that both sennoside A and sennoside B increased prostaglandin E2 (PGE2) levels and inhibited H(+)/K(+)-ATPase (proton pump). In a rat model, both compounds reduced gastric juice, total acidity and increased pH, indicating that proton pump inhibition reduces gastric acid secretion. Furthermore, sennoside A and B increased PGE2 in a concentration-dependent manner. In a gastric emptying and intestinal transporting rate experiment, both sennoside A and sennoside B accelerated motility. Our results thus suggest that sennoside A and sennoside B possess significant gastroprotective activities and they might be useful for the treatment of gastric disease.
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, Vetec, reagent grade, 99%