  • Search Results
  • Protein adsorption, desorption, and aggregation mediated by solid-liquid interfaces.
Protein adsorption, desorption, and aggregation mediated by solid-liquid interfaces.
Journal of pharmaceutical sciences (2015-04-08)
Tatiana Perevozchikova, Hirsh Nanda, Douglas P Nesta, Christopher J Roberts
ABSTRACT
Adsorption of proteins to solid-fluid interfaces is often empirically found to promote formation of soluble aggregates and larger, subvisible, and visible particles, but key stages in this process are often difficult to probe directly. Aggregation mediated by adsorption to water-silicon oxide (SiOx) interfaces, akin to hydrated glass surfaces, was characterized as a function of pH and ionic strength for alpha-chymotrypsinogen (aCgn) and for a monoclonal antibody (IgG1). A flow cell permitted neutron reflectivity for protein layers adsorbed to clean SiOx surfaces, as well as after successive "rinse" steps. Aggregates recovered in solution after gently "rinsing" the surface were characterized by neutron scattering, microscopy, and fluorescence spectroscopy. IgG1 molecules oriented primarily "flat" against the SiOx surface, with the primary protein layer desorbed to a minimal extent, whereas a diffuse overlayer was easily rinsed off. aCgn molecules were resistant to desorption when they appeared to be unfolded at the interface, but were otherwise easily removed. For cases where strong binding occurred, protein that did desorb was a mixture of monomer and small amounts of HMW aggregates (for aCgn) or subvisible particles (for IgG1). Changes in adsorption and/or unfolding with pH indicated that electrostatic interactions were important in all cases.
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Citric acid, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, Vetec, reagent grade, 98%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%