  • Search Results
  • Grafted-double walled carbon nanotubes as electrochemical platforms for immobilization of antibodies using a metallic-complex chelating polymer: Application to the determination of adiponectin cytokine in serum.
Grafted-double walled carbon nanotubes as electrochemical platforms for immobilization of antibodies using a metallic-complex chelating polymer: Application to the determination of adiponectin cytokine in serum.
Biosensors & bioelectronics (2015-06-21)
Irene Ojeda, Myriam Barrejón, Luis M Arellano, Araceli González-Cortés, Paloma Yáñez-Sedeño, Fernando Langa, José M Pingarrón
ABSTRACT
An electrochemical immunosensor for adiponectin (APN) using screen printed carbon electrodes (SPCEs) modified with functionalized double-walled carbon nanotubes (DWCNTs) as platforms for immobilization of the specific antibodies is reported. DWCNTs were functionalized by treatment with 4-aminobenzoic acid (HOOC-Phe) in the presence of isoamylnitrite resulting in the formation of 4-carboxyphenyl-DWCNTs. The oriented binding of specific antibodies toward adiponectin was accomplished by using the metallic-complex chelating polymer Mix&Go™. The HOOC-Phe-DWCNTs-modified SPCEs were characterized by cyclic voltammetry and compared with HOOC-Phe-SWCNTs/SPCE. The different variables affecting the performance of the developed immunosensor were optimized. Under the selected conditions, a calibration plot for APN was constructed showing a range of linearity extending between 0.05 and 10.0 μg/mL which is adequate for the determination of the cytokine in real samples. A detection limit of 14.5 ng/mL was achieved. The so prepared immunosensor exhibited a good reproducibility for the APN measurements, excellent storage stability and selectivity, and a much shorter assay time than the available ELISA kits. The usefulness of the immunosensor for the analysis of real samples was demonstrated by analyzing human serum from female or male healthy patients.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, 99.998% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate monobasic, Vetec, reagent grade, 99%