  • Search Results
  • Monosodium urate crystals trigger Nrf2- and heme oxygenase-1-dependent inflammation in THP-1 cells.
Monosodium urate crystals trigger Nrf2- and heme oxygenase-1-dependent inflammation in THP-1 cells.
Cellular & molecular immunology (2014-08-12)
Jhih-Jia Jhang, Yu-Ting Cheng, Cheng-Ying Ho, Gow-Chin Yen
ABSTRACT
Gouty arthritis is an inflammatory disease that is caused by an accumulation of monosodium urate (MSU) crystals in the joints. MSU is capable of activating the nucleotide-binding oligomerization domain-like receptor pyrin domain containing 3 (NLRP3) inflammasome, leading to interleukin-1β (IL-1β) secretion. Reactive oxygen species (ROS) are major mediators of the NLRP3/IL-1β interaction. Although nuclear factor E2-related factor 2 (Nrf2) is recognized as a transcription factor that is involved in the response to oxidative stress, the effect of MSU on Nrf2 and on Nrf2-mediated antioxidant enzymes remains unclear. The treatment of THP-1 monocytes using phorbol 12-myristate 13-acetate (PMA) was shown to initiate inflammatory responses. Here, we showed that THP-1 cells, following treatment with MSU crystals, significantly increased IL-1β release, NLRP3 inflammasome activation and ROS production. MSU also promoted the nuclear translocation of Nrf2 and activated lysosomal destabilization. Moreover, the levels of heme oxygenase-1 (HO-1) in gene and protein expressions were upregulated by MSU. MSU-induced IL-1β secretion and NLRP3 inflammasome activation were inhibited by the knockdown of Nrf2 and via the HO-1 inhibitor zinc (II) protoporphyrin IX (ZnPP). In addition, HO-1 inhibition increased the level of superoxide anion production and the consumption of glutathione. These findings suggest that Nrf2 and HO-1 mediate redox homeostasis and interact with pro-inflammatory factors in MSU-challenged THP-1 cells, thereby providing new insight into how MSU-induced gouty inflammation is mediated by specific mechanisms that are involved in the Nrf2/Ho-1 antioxidant signaling pathway.Cellular & Molecular Immunology advance online publication, 11 August 2014; doi:10.1038/cmi.2014.65.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Protoporphyrin IX, ≥95%
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
SAFC
HEPES
Sigma-Aldrich
Magnesium chloride solution, 0.1 M