  • Search Results
  • Effects of the polyphenol resveratrol on contractility of human term pregnant myometrium.
Effects of the polyphenol resveratrol on contractility of human term pregnant myometrium.
Molecular human reproduction (2015-03-10)
R Novaković, N Radunović, J Marković-Lipkovski, S Ćirović, B Beleslin-Čokić, B Ilić, B Ivković, H Heinle, V Živanović, L J Gojković-Bukarica
ABSTRACT
The ideal agent for prevention and treatment of uterine abnormal contractility has not been found. The polyphenol resveratrol possesses a wide spectrum of pharmacologic properties, but its influence on the contractility of human myometrium is not defined. The present study evaluated the effect of resveratrol on the oxytocin-induced contractions of human term pregnant myometrium in vitro and the contribution of different K(+) channels to resveratrol action. Resveratrol induced a concentration-dependent relaxation of myometrium contractions (pD2 value and maximal responses were 4.52 and 82.25%, respectively). Glibenclamide, a selective blocker of ATP-sensitive (KATP), iberiotoxin, a selective blockers of big-calcium sensitive (BK(Ca)) and 4-aminopiridine, a non-selective blocker of voltage-sensitive (Kv) channels induced a significant shift to the right of the concentration-response curves of resveratrol. Inhibition achieved by 0.1 mM resveratrol was insensitive to all K(+) channel blockers. A K(+) channel opener, pinacidil, inhibited oxytocin-induced contractions of pregnant myometrium with comparable potency and efficacy to resveratrol (pD2 values and maximal relaxation were 4.52 and 83.67%, respectively). Based on K(+) channel opener/blocker affinities, it appears that the inhibitory response of resveratrol involves different myometrial K(+) channels. When applied in high concentrations, resveratrol has an additional K(+)-channel-independent mechanism(s) of action. Furthermore, immunohistochemistry staining and western blot analyses detected the presence and distribution of KATP, BK(Ca) and Kv channel proteins in pregnant myometrium.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Supelco
Hematoxylin
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Lithium dodecyl sulfate, Vetec, reagent grade, ≥98.5%
Sigma-Aldrich
Sodium deoxycholate, Vetec, reagent grade, ≥97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)