  • Search Results
  • Expression and activity of the Calvin-Benson-Bassham cycle transcriptional regulator CbbR from Acidithiobacillus ferrooxidans in Ralstonia eutropha.
Expression and activity of the Calvin-Benson-Bassham cycle transcriptional regulator CbbR from Acidithiobacillus ferrooxidans in Ralstonia eutropha.
FEMS microbiology letters (2015-07-15)
Mario Esparza, Eugenia Jedlicki, Mark Dopson, David S Holmes
ABSTRACT
Autotrophic fixation of carbon dioxide into cellular carbon occurs via several pathways but quantitatively, the Calvin-Benson-Bassham cycle is the most important. CbbR regulates the expression of the cbb genes involved in CO2 fixation via the Calvin-Benson-Bassham cycle in a number of autotrophic bacteria. A gene potentially encoding CbbR (cbbR(AF)) has been predicted in the genome of the chemolithoautotrophic, extreme acidophile Acidithiobacillus ferrooxidans. However, this microorganism is recalcitrant to genetic manipulation impeding the experimental validation of bioinformatic predictions. Two novel functional assays were devised to advance our understanding of cbbR(AF) function using the mutated facultative autotroph Ralstonia eutropha H14 ΔcbbR as a surrogate host to test gene function: (i) cbbR(AF) was expressed in R. eutropha and was able to complement ΔcbbR; and (ii) CbbR(AF) was able to regulate the in vivo activity of four A. ferrooxidans cbb operon promoters in R. eutropha. These results open up the use of R. eutropha as a surrogate host to explore cbbR(AF) activity.
Product Description
Adapter for ACE Micro/Mini-Lab® kit, Vacuum-takeoff with hose barb, joint: ST/NS 14/10
Adapter for ACE Micro/Mini-Lab® kit, Mini adapter
Adapter for ACE Micro/Mini-Lab® kit, Distilling, joint: ST/NS 14/10
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 3.6 gal (13.6 L)
Mini chemical spill kit
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 1.8 gal (6.8 L)