  • Search Results
  • EPA-enriched phospholipids ameliorate cancer-associated cachexia mainly via inhibiting lipolysis.
EPA-enriched phospholipids ameliorate cancer-associated cachexia mainly via inhibiting lipolysis.
Food & function (2015-09-17)
Lei Du, Yu-Hong Yang, Yu-Ming Wang, Chang-Hu Xue, Hideyuki Kurihara, Koretaro Takahashi
ABSTRACT
Excessive loss of fat mass is considered as a key feature of body weight loss in cancer-associated cachexia (CAC). It affects the efficacy and tolerability of cancer therapy and reduces the quality and length of cancer patients' lives. The aim of the present study was to evaluate the effects of EPA-enriched phospholipids (EPA-PL) derived from starfish Asterias amurensis on cachectic weight loss in mice bearing S180 ascitic tumor, and TNF-α-stimulated lipolysis in 3T3-L1 adipocytes and to elucidate the possible mechanisms involved. Our findings revealed that oral administration of EPA-PL at 100 mg per kg body weight (BW) per day for 14 days prevented body weight loss in CAC mice by preserving the white adipose tissue (WAT) mass. We found that serum levels of nonesterified fatty acid (NEFA) and pro-inflammatory cytokines such as tumor necrosis factor α (TNF-α) and interleukin (IL)-6 increased in CAC mice but decreased significantly after oral treatment of EPA-PL. In addition, EPA-PL treatment also suppressed the overexpression of several key lipolytic factors and raised the mRNA levels of some adipogenic factors in the WAT of CAC mice. Moreover, treatment of EPA-PL (200 and 400 μM) markedly inhibited TNF-α-stimulated lipolysis in adipocytes. Furthermore, the antilipolytic effects of EPA-PL were stimulated by the extracellular signal-regulated kinase 1/2 (ERK 1/2) inhibitor PD 98059 and blocked via the AMP-activated protein kinase (AMPK) inhibitor compound C and the phosphoinositide-3-kinase (PI3K) inhibitor LY 294002. Taken together, these data suggest that the dietary EPA-PL ameliorates CAC mainly via inhibiting lipolysis and at least in part for recovering the function of adipogenesis.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Glycerol solution, puriss. p.a., 86-89% (T)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)