  • Search Results
  • Cyclosporin A-sensitive cytotoxicity of flurbiprofen non-stereoselectively mediated by cytochrome P450 metabolism in three-dimensional cultured rat hepatocytes.
Cyclosporin A-sensitive cytotoxicity of flurbiprofen non-stereoselectively mediated by cytochrome P450 metabolism in three-dimensional cultured rat hepatocytes.
The Journal of pharmacy and pharmacology (2015-06-24)
Tadatoshi Tanino, Yoshinori Funakami, Noriaki Nagai, Yoshihisa Kato
ABSTRACT
2-Arylpropionic acid (profen) drugs are associated with severe hepatotoxicity; however, risk factors are still poorly understood. Acyl-coenzyme A (acyl-CoA) thioesters of profen drugs play a more important role in the covalent binding to rat hepatocyte proteins than the respective acyl-glucuronides. Therefore, we examined whether acyl-glucuronides, acyl-CoA thioesters and oxidative metabolites of profen drugs stereoselectively participated in liver damage. Cytotoxicity was determined by measuring lactate dehydrogenase (LDH) leakage from three-dimensional cultured rat hepatocytes. LDH leakage was not induced by R-2-phenylpropionic acid and R-ibuprofen greatly forming acyl-CoA thioesters. S-Naproxen metabolized mainly by Uridine 5'-diphosphate (UDP)-glucuronosyl-transferase did not enhance LDH leakage. However, flurbiprofen (FLP) induced LDH leakage. A selective cytochrome P450 (CYP) 2C11 inhibitor suppressed 40-50% of the R-FLP and S-FLP-induced cytotoxicity. Borneol non-stereoselectively accelerated the FLP-induced cytotoxicity. The R-FLP-induced cytotoxicity decreased intracellular adenosine triphosphate (ATP) levels to 50% of untreated hepatocytes. An inhibitor of mitochondrial permeability transition pore, cyclosporin A (Cys A), rescued ATP levels and LDH leakage back to control levels. The reactive acyl-CoA thioesters and acyl-glucuronides were not associated with liver damage, denying one of the leading hypotheses. CYP metabolism of FLP non-stereoselectively participated in Cys A-sensitive cytotoxicity, suggesting mitochondrial injury.
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
DIC, 99%
Sigma-Aldrich
4-Hydroxy-TEMPO, 97%
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
2-Fluorobiphenyl, 96%
Sigma-Aldrich
1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid, 98%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
(±)-CPP, solid
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in dichloromethane
Sigma-Aldrich
Sulfaphenazole, ≥98% (HPLC)
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in THF