  • Search Results
  • Visualization of Lipid Membrane Reorganization Induced by a Pore-Forming Toxin Using High-Speed Atomic Force Microscopy.
Visualization of Lipid Membrane Reorganization Induced by a Pore-Forming Toxin Using High-Speed Atomic Force Microscopy.
ACS nano (2015-07-30)
Neval Yilmaz, Toshihide Kobayashi
ABSTRACT
We examined the effect of a sphingomyelin (SM)-binding pore-forming toxin (PFT), lysenin, on the dynamics of a phase-separated membrane of SM, where SM formed liquid-ordered (Lo) domains with cholesterol (Chol) within a phosphatidylcholine-rich liquid-disordered (Ld) phase. We visualized the lysenin-induced membrane reorganization using high-speed atomic force microscope (HS-AFM). Lysenin oligomerized on the SM-rich Lo domain and simultaneously its oligomers assembled into a hexagonal close-packed (hcp) structure. The phase boundary was stable during the assembling of lysenin on the SM-rich domain, indicating that lysenin did not affect the line tension between Lo and Ld phases. After the full coverage of the SM-rich domain by oligomers, their hcp assembly gradually expanded into the Ld phase and eventually covered the entire membrane. Our results suggest that pore formation, i.e., insertion of lysenin into the membrane in its oligomeric state, induced the exclusion of SM and Chol from the SM-rich domain, which was followed by further binding and oligomerization of lysenin.
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder