  • Search Results
  • In vivo prediction of goat kids body composition from the deuterium oxide dilution space determined by isotope-ratio mass spectrometry.
In vivo prediction of goat kids body composition from the deuterium oxide dilution space determined by isotope-ratio mass spectrometry.
Journal of animal science (2015-10-07)
S Lerch, M L Lastel, C Grandclaudon, C Brechet, G Rychen, C Feidt
ABSTRACT
Deuterium oxide dilution space (DOS) determination is one of the most accurate methods for in vivo estimation of ruminant body composition. However, the time-consuming vacuum sublimation of blood preceding infrared spectroscopy analysis, which is traditionally used to determine deuterium oxide (DO) concentration, limits its current use. The use of isotope-ratio mass spectrometry (IRMS) to determine the deuterium enrichment and thus quantify DO in plasma could counteract this limitation by reducing the sample preparation for plasma deproteinisation through centrifugal filters. The aim of this study was to validate the DOS technique using IRMS in growing goat kids to establish in vivo prediction equations of body composition. Seventeen weaned male Alpine goat kids (8.6 wk old) received a hay-based diet supplemented with 2 types of concentrates providing medium ( = 9) or high ( = 8) energy levels. Kids were slaughtered at 14.0 ( = 1, medium-energy diet), 17.2 ( = 4, medium-energy diet, and = 4, high-energy diet), or 21.2 wk of age ( = 4, medium-energy diet, and = 4, high-energy diet). Two days before slaughter, DOS was determined after an intravenous injection of 0.2 g DO/kg body mass (BM) and the resulting study of DO dilution kinetics from 4 plasma samples (+5, +7, +29, and +31 h after injection). The deuterium enrichment was analyzed by IRMS. After slaughter, the gut contents were discarded, the empty body (EB) was minced, and EB water, lipid, protein, ash, and energy contents were measured by chemical analyses. Prediction equations for body components measured postmortem were computed from in vivo BM and DOS. The lack of postmortem variation of fat-free EB composition was confirmed (mean of 75.3% [SD 0.6] of water), and the proportion of lipids in the EB tended ( = 0.06) to be greater for the high-energy diet (13.1%) than for the medium-energy diet (11.1%). There was a close negative relationship (residual CV [rCV] = 3.9%, = 0.957) between EB water and lipid content, whereas DOS was closely related to total body water (rCV = 2.9%, = 0.944) but DOS overestimated it by 5.8%. Adding DOS to BM improved the in vivo predictions of EB lipid and energy content (rCV = 13.1% and rCV = 7.9%, respectively) but not those of protein or ash. Accuracy of the obtained prediction equations was similar to those reported in studies determining DOS by infrared spectroscopy. Therefore, the use of IRMS to quantify DOS provides a highly accurate measure of the in vivo body composition in goat kids.
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture