  • Search Results
  • Fatty acid composition analysis in polysorbate 80 with high performance liquid chromatography coupled to charged aerosol detection.
Fatty acid composition analysis in polysorbate 80 with high performance liquid chromatography coupled to charged aerosol detection.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-03)
David Ilko, Alexandra Braun, Oliver Germershaus, Lorenz Meinel, Ulrike Holzgrabe
ABSTRACT
The fatty acid (FA) composition of polysorbate 80 (PS80), a sorbitan oleic acid ester copolymerized with about 20mole of ethylene oxide, is typically characterized by gas chromatography. Here, an alternative method was developed. After saponification with potassium hydroxide the FA fraction was collected with liquid-liquid extraction using methyl-tert-butyl ether. HPLC in combination with a Corona® charged aerosol detector (CAD) was applied for the separation and detection. The method was fully validated in terms of specificity, repeatability, limits of quantification, linearity, range, accuracy and robustness. The characterization of 16 different PS80 batches demonstrated variability regarding their FA composition, with e.g. the amount of oleic acid ranging from 67.8±0.7% to 96.6±1.4%. Furthermore, we identified petroselinic acid, a double-bond positional isomer to oleic acid in all batches, an FA not known to pharmacopoeias at present. In addition, 11-hydroxy-9-octadecenoic acid, an oxidation product of oleic acid was identified. Structure elucidation was performed by means of HPLC-MS/MS. In addition, the method was expanded to the evaluation of the free FAs. Having determined the entire FA composition, the acid value according to EP and USP can be calculated.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Heptadecanoic acid, ≥98%
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG