  • Search Results
  • Chronic exposure to pollutants in Madín Reservoir (Mexico) alters oxidative stress status and flesh quality in the common carp Cyprinus carpio.
Chronic exposure to pollutants in Madín Reservoir (Mexico) alters oxidative stress status and flesh quality in the common carp Cyprinus carpio.
Environmental science and pollution research international (2015-01-15)
Gabriela Morachis-Valdez, Octavio Dublán-García, Leticia Xochitl López-Martínez, Marcela Galar-Martínez, Karinne Saucedo-Vence, Leobardo Manuel Gómez-Oliván
ABSTRACT
Madín Reservoir (MR) is located on the Río Tlalnepantla in Mexico. Previous studies seeking to identify pollutants at this site evidence that MR water contains a considerable metal load as well as nonsteroidal anti-inflammatory drugs (NSAIDs) at concentrations above those determined suitable for aquatic life. This study aimed to evaluate whether chronic exposure to pollutants in MR alters oxidative stress status and flesh quality in muscle of the common carp Cyprinus carpio. The following biomarkers were evaluated in muscle of carp caught in the general area of discharge from the town of Viejo Madín: hydroperoxide content (HPC), lipid peroxidation (LPX), protein carbonyl content (PCC), and activity of the antioxidant enzymes superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase (GPx). Physicochemical and textural properties of muscle were also evaluated. Results show that the metals Al and Fe were accumulated in muscle of C. carpio at levels of 21.3 and 29.6 μg L(-1), respectively, and the NSAIDs diclofenac, ibuprofen, and naproxen at levels from 0.08 to 0.21 ng L(-1). Fish exposed to discharge from the town of Viejo Madín showed significant increases in HPC (9.77 %), LPX (69.33 %), and PCC (220 %) with respect to control specimens (p < 0.05). Similarly, enzyme activity increased significantly: SOD (80.82 %), CAT (98.03 %), and GPx (49.76 %). In muscle, physicochemical properties evidenced mainly significant reductions compared to control values while textural properties showed significant increases. Thus, water in this reservoir is contaminated with xenobiotics that alter some biological functions in C. carpio, a fish species consumed by the local human population.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Potassium phosphate dibasic, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M