  • Search Results
  • Comparison of in-cell lipid removal efficiency of adsorbent mixtures for extraction of polybrominated diphenyl ethers in fish.
Comparison of in-cell lipid removal efficiency of adsorbent mixtures for extraction of polybrominated diphenyl ethers in fish.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-04-02)
Peter Wardrop, Paul D Morrison, Jeff G Hughes, Bradley O Clarke
ABSTRACT
Polybrominated diphenyl ethers (PBDEs) have become ubiquitous environmental contaminants due to their incorporation into many consumer products. Their ability to bioaccumulate to alarming levels in fat-rich matrices such as fish demands fast and efficient methods to monitor these contaminants. We present an analytical method for selective-pressurised liquid extraction (S-PLE) of PBDEs from fish tissue. Fat removal performance of different mixtures of Florisil, silica gel and sulphuric acid-impregnated silica gel were evaluated using a response surface experimental design approach for determining the optimal fat-retaining mixture for S-PLE. Acid-silica gel had the greatest individual effect on fat retention; with a two-thirds acid-silica one-third Florisil mixture found to be the most efficient (>97%). Method validation was performed using recovery experiments at three spiked concentration levels (0.05, 0.5 and 5ngg(-1) ww). Mean recoveries of target analytes in spiked samples ranged from 70 to 124%, with relative standard deviations <27%. The S-PLE lipid removal efficiency combined with the sensitivity of triple quadrupole mass spectrometers provides a fast and comparatively inexpensive analytical method for analysis of PBDEs in fish samples.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%