  • Search Results
  • Enhanced oxidative stress sensitizes the mitochondrial permeability transition pore to opening in heart from Zucker Fa/fa rats with type 2 diabetes.
Enhanced oxidative stress sensitizes the mitochondrial permeability transition pore to opening in heart from Zucker Fa/fa rats with type 2 diabetes.
Life sciences (2015-09-27)
Adriana Riojas-Hernández, Judith Bernal-Ramírez, David Rodríguez-Mier, Flor E Morales-Marroquín, Elvia M Domínguez-Barragán, Cuauhtémoc Borja-Villa, Irais Rivera-Álvarez, Gerardo García-Rivas, Julio Altamirano, Noemí García
ABSTRACT
Obesity and diabetes mellitus type 2 (DM2) frequently coexist and increase the propensity of cardiovascular dysfunction by numerous mechanisms. Chief among them are oxidative stress and Ca(2+) dysregulation, and both are inducers of the mitochondrial permeability transition pore (MPTP). Nevertheless, it is unknown whether MPTP formation is triggered in DM2 animals, and thereby contributing to cardiac dysfunction. We assessed MPTP sensitivity and reactive oxygen species production in cardiac mitochondria, as well as cytosolic Ca(2+) handling in ventricular myocytes from rats with DM2. Male Zucker Fa/fa rats (Fa/fa) 32weeks old presenting DM2, concentric hypertrophy, and diastolic dysfunction were used. MPTP formation was evaluated in isolated mitochondria and Ca(2+) handling in ventricular myocytes, by spectrophotometric and confocal microscope techniques, respectively. We found that the systolic Ca(2+) transient relaxation was ~40% slower, while mitochondrial H2O2 production increased by ~6-fold. MPTP opening in isolated mitochondria from Fa/fa (mFa/fa) was more sensitive to Ca(2+) than in mitochondria from lean rats (mLean), and correlated with increased thiol group exposure. The mFa/fa showed decreased oxidative phosphorylation capacity. The ATP content decreased in myocytes, while the PCr/ATP ratio remained unchanged and caspase 9 activity largely increased in myocytes from Fa/fa animals. Our results showed that oxidative stress and diastolic Ca(2+) dysregulation increased MPTP sensitivity leading to mitochondrial dysfunction and apoptosis. Mitochondrial dysfunction could compromise ATP synthesis, and lower ATP could be linked to decreased SERCA2 activity, which might underlie diastolic dysfunction. Prolonged Ca(2+) transients might further exacerbate mitochondrial dysfunction.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, Vetec, reagent grade, 99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)