  • Search Results
  • Nicking endonuclease-assisted signal amplification of a split molecular aptamer beacon for biomolecule detection using graphene oxide as a sensing platform.
Nicking endonuclease-assisted signal amplification of a split molecular aptamer beacon for biomolecule detection using graphene oxide as a sensing platform.
The Analyst (2015-10-27)
Xiang Li, Xuelian Ding, Jing Fan
ABSTRACT
Sensitive and selective detection of ultralow concentrations of specific biomolecules is important in early clinical diagnoses and biomedical applications. Many types of aptasensors have been developed for the detection of various biomolecules, but usually suffer from false positive signals and high background signals. In this work, we have developed an amplified fluorescence aptasensor platform for ultrasensitive biomolecule detection based on enzyme-assisted target-recycling signal amplification and graphene oxide. By using a split molecular aptamer beacon and a nicking enzyme, the typical problem of false positive signals can be effectively resolved. Only in the presence of a target biomolecule, the sensor system is able to generate a positive signal, which significantly improves the selectivity of the aptasensor. Moreover, using graphene oxide as a super-quencher can effectively reduce the high background signal of a sensing platform. We select vascular endothelial growth factor (VEGF) and adenosine triphosphate (ATP) as model analytes in the current proof-of-concept experiments. It is shown that under optimized conditions, our strategy exhibits high sensitivity and selectivity for the quantification of VEGF and ATP with a low detection limit (1 pM and 4 nM, respectively). In addition, this biosensor has been successfully utilized in the analysis of real biological samples.
Sigma-Aldrich
Lactic acid solution, Vetec, reagent grade, 85%
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl