  • Search Results
  • Development of an LC-MS/MS assay for the quantitative determination of the intracellular 5-fluorouracil nucleotides responsible for the anticancer effect of 5-fluorouracil.
Development of an LC-MS/MS assay for the quantitative determination of the intracellular 5-fluorouracil nucleotides responsible for the anticancer effect of 5-fluorouracil.
Journal of pharmaceutical and biomedical analysis (2015-03-26)
Ellen J B Derissen, Michel J X Hillebrand, Hilde Rosing, Jan H M Schellens, Jos H Beijnen
ABSTRACT
5-Fluorouracil (5-FU) and its oral prodrug capecitabine are among the most widely used chemotherapeutics. For cytotoxic activity, 5-FU requires cellular uptake and intracellular metabolic activation. Three intracellular formed metabolites are responsible for the antineoplastic effect of 5-FU: 5-fluorouridine 5'-triphosphate (FUTP), 5-fluoro-2'-deoxyuridine 5'-triphosphate (FdUTP) and 5-fluoro-2'-deoxyuridine 5'-monophosphate (FdUMP). In this paper, we describe the development of an LC-MS/MS assay for quantification of these active 5-FU nucleotides in peripheral blood mononuclear cells (PBMCs). Because the intracellular 5-FU nucleotide concentrations were very low, maximization of the release from the cell matrix and minimization of interference were critical factors. Therefore, a series of experiments was performed to select the best method for cell lysis and nucleotide extraction. Chromatography was optimized to obtain separation from endogenous nucleotides, and the effect of different cell numbers was examined. The assay was validated for the following concentration ranges in PBMC lysate: 0.488-19.9 nM for FUTP, 1.66-67.7 nM for FdUTP and 0.748-30.7 nM for FdUMP. Accuracies were between -2.2 and 7.0% deviation for all analytes, and the coefficient of variation values were ≤ 4.9%. The assay was successfully applied to quantify 5-FU nucleotides in PBMC samples from patients treated with capecitabine and patients receiving 5-FU intravenously. FUTP amounts up to 3054 fmol/10(6) PBMCs and FdUMP levels up to 169 fmol/10(6) PBMCs were measured. The FdUTP concentrations were below the lower limit of quantification. To our knowledge, this is the first time that 5-FU nucleotides were quantified in cells from patients treated with 5-FU or capecitabine without using a radiolabel.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Uridine, Vetec, reagent grade, 99%
Sigma-Aldrich
Ammonium acetate, Vetec, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
5-Fluorouridine, proapoptotic anitproliferative plant growth regulator
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.