  • Search Results
  • Studies of air, water, and ethanol vapor atmospheric pressure plasmas for antimicrobial applications.
Studies of air, water, and ethanol vapor atmospheric pressure plasmas for antimicrobial applications.
Biointerphases (2015-03-27)
James R Ferrell, Erinn R Bogovich, Nicholas R Lee, Robert L Gray, Daphne D Pappas
ABSTRACT
The generation of air-based plasmas under atmospheric plasma conditions was studied to assess their antimicrobial efficacy against commonly found pathogenic bacteria. The mixture of initial gases supplied to the plasma was found to be critical for the formation of bactericidal actives. The optimal gas ratio for bactericidal effect was determined to be 99% nitrogen and 1% oxygen, which led to a 99.999% reduction of a pathogenic strain of Escherichia coli on stainless steel surfaces. The experimental substrate, soil load on the substrate, flow rate of the gases, and addition of ethanol vapor all were found to affect antimicrobial efficacy of studied plasmas. Optical emission spectroscopy was used to identify the species that were present in the plasma bulk phase for multiple concentrations of nitrogen and oxygen ratios. The collected spectra indicate a unique series of bands present in the ultraviolet region of the electromagnetic spectrum that can be attributed to nitric oxide species known to be highly antimicrobial. This intense spectral profile dramatically changes as the concentration of nitrogen decreases.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis