  • Search Results
  • Contrasting Nutrient Mitigation and Denitrification Potential of Agricultural Drainage Environments with Different Emergent Aquatic Macrophytes.
Contrasting Nutrient Mitigation and Denitrification Potential of Agricultural Drainage Environments with Different Emergent Aquatic Macrophytes.
Journal of environmental quality (2015-10-06)
Jason M Taylor, Matthew T Moore, J Thad Scott
ABSTRACT
Remediation of excess nitrogen (N) in agricultural runoff can be enhanced by establishing wetland vegetation, but the role of denitrification in N removal is not well understood in drainage ditches. We quantified differences in N retention during experimental runoff events followed by stagnant periods in mesocosms planted in three different vegetation treatments: unvegetated, cutgrass [ (L.) Sw.], and common cattail ( L.). We also quantified denitrification rates using membrane inlet mass spectrometry from intact cores extracted from each mesocosm treatment. All treatments retained 60% or more of NO-N loads during the 6-h experimental runoff event, but mesocosms planted with cutgrass had significantly higher (68%) retention than the cattail (60%) or unvegetated (61%) treatments. After the runoff event, mesocosms planted in cattail reduced NO-N concentrations by >95% within 24 h and cutgrass achieved similar reductions within 48 h, whereas reductions in the unvegetated mesocosms were significantly less (65%). Cores from cutgrass mesocosms had significantly higher average denitrification rates (5.93 mg m h), accounting for as much as 56% of the immobilized NO-N within 48 h, whereas denitrification rates were minimal in cores from the unvegetated (-0.19 mg m h) and cattail (0.2 mg m h) mesocosms. Our findings have implications for mitigating excess NO-N in agricultural runoff. While vegetated treatments removed excess NO-N from the water column at similar and significantly higher rates than unvegetated treatments, the high denitrification rates observed for cutgrass highlight the potential for permanent removal of excess N from agricultural runoff in vegetated ditches and wetlands.
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N