  • Search Results
  • In-coupled syringe assisted octanol-water partition microextraction coupled with high-performance liquid chromatography for simultaneous determination of neonicotinoid insecticide residues in honey.
In-coupled syringe assisted octanol-water partition microextraction coupled with high-performance liquid chromatography for simultaneous determination of neonicotinoid insecticide residues in honey.
Talanta (2015-04-18)
Jitlada Vichapong, Rodjana Burakham, Supalax Srijaranai
ABSTRACT
A simple and fast method namely in-coupled syringe assisted octanol-water partition microextraction combined with high performance liquid chromatography (HPLC) has been developed for the extraction, preconcentration and determination of neonicotinoid insecticide residues (e.g. imidacloprid, acetamiprid, clothianidin, thiacloprid, thiamethoxam, dinotefuran, and nitenpyram) in honey. The experimental parameters affected the extraction efficiency, including kind and concentration of salt, kind of disperser solvent and its volume, kind of extraction solvent and its volume, shooting times and extraction time were investigated. The extraction process was carried out by rapid shooting of two syringes. Therefore, rapid dispersion and mass transfer processes was created between phases, and thus affects the extraction efficiency of the proposed method. The optimum extraction conditions were 10.00 mL of aqueous sample, 10% (w/v) Na2SO4, 1-octanol (100µL) as an extraction solvent, shooting 4 times and extraction time 2min. No disperser solvent and centrifugation step was necessary. Linearity was obtained within the range of 0.1-3000 ngmL(-1), with the correlation coefficients greater than 0.99. The high enrichment factor of the target analytes was 100 fold and low limit of detection (0.25-0.50 ngmL(-1)) could be obtained. This proposed method has been successfully applied in the analysis of neonicotinoid residues in honey, and good recoveries in the range of 96.93-107.70% were obtained.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), Customized plastic drum
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), One-time steel-plastic (SP) drum
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ultrapure Acetonitrile
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
1-Octanol, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium sulfate, Vetec, reagent grade, 99%