  • Search Results
  • Synthesis, molecular docking and biological evaluation of 3-arylfuran-2(5H)-ones as anti-gastric ulcer agent.
Synthesis, molecular docking and biological evaluation of 3-arylfuran-2(5H)-ones as anti-gastric ulcer agent.
Bioorganic & medicinal chemistry (2015-06-07)
Xu-Dong Wang, Wei Wei, Peng-Fei Wang, Li-Cheng Yi, Wei-Kang Shi, Yong-Xiang Xie, Lang-Zhou Wu, Nian Tang, Liang-Song Zhu, Jia Peng, Chan Liu, Xian-Hui Li, Shi Tang, Zhu-Ping Xiao, Hai-Liang Zhu
ABSTRACT
3-Arylfuran-2(5H)-one derivatives show good antibacterial activity and were determined as tyrosyl-tRNA synthetase (TyrRS) inhibitors. In a systematic medicinal chemistry exploration, we demonstrated chemical opportunities to treat infections caused by Helicobacter pylori. Twenty 3-arylfuran-2(5H)-ones were synthesized and evaluated for anti-H. pylori, antioxidant and anti-urease activities which are closely interconnected with H. pylori infection. The results displayed that some of the compounds show excellent antioxidant activity, and good anti-H. pylori and urease inhibitory activities. Out of these compounds, 3-(3-methylphenyl)furan-2(5H)-one (b9) showed the most potent antioxidant activity (IC50=8.2 μM) and good anti-H. pylori activity (MIC50=2.6 μg/mL), and it can be used as a good candidate for discovering novel anti-gastric ulcer agent.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Amoxicillin, potency: ≥900 μg per mg
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
SAFC
L-Tyrosine
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D