  • Search Results
  • Kudoa saudiensis sp. n. (Myxosporea: Multivalvulida) infecting oocytes of the Indian mackerel Rastrelliger kanagurta (Perciformes: Scombridae).
Kudoa saudiensis sp. n. (Myxosporea: Multivalvulida) infecting oocytes of the Indian mackerel Rastrelliger kanagurta (Perciformes: Scombridae).
Folia parasitologica (2015-05-12)
Lamjed Mansour, Abdel Halim Harrath, Abdel-Azeem S Abdel-Baki, Saleh Al-Quraishy, Suliman Y Omar Al
ABSTRACT
During a survey the occurrence of Kudoa quraishii Mansour, Harrath, Abd-Elkader, Alwasel, Abdel-Baki et Al Omar, 2014, recently identified in the muscles of the Indian mackerel, Rastrelliger kanagurta (Cuvier), a species of Kudoa Meglitsch, 1947 infecting oocytes of mature females of the same host fish was found. The new species, for which the name Kudoa saudiensis sp. n. is proposed, infects oocytes that are enlarged with a whitish colour. The parasite develops in vesicular polysporous plasmodia within the oocyte. Infection occurs with a mean prevalence of 20% (7/35) of examined females. Mature spores are quadratic in shape in apical view, having four equal valves and four symmetrical polar capsules. Fresh spores are 2.4-3.6 µm long (mean ± SD 3.1 ± 0.3 µm), 4.3-5.4 µm (4.7 ± 0.3 µm) wide and 3.4-4.3 µm (3.8 ± 0.3 µm) in thickness and long. The smaller size of the new Kudoa species was the distinctive feature that separates it from all previously described species. Molecular analysis based on the SSU rDNA sequences shows that the highest percentage of similarity of 98.5% was observed with K. ovivora Swearer et Robertson, 1999, reported from oocytes of labroid fish from the Caribbean coasts of Panama. The percentage of similarity was 98% with K. azevedoi Mansour, Thabet, Chourabi, Harrath, Gtari, Al Omar et Ben Hassine, 2013 and 89% with K. quraishii. Phylogenetic analysis of the SSU and LSU rDNA data revealed a consistent of the new species with K. azevedoi and K. ovivora. Our findings support the creation of Kudoa saudiensis sp. n. that infects oocytes of the Indian mackerel Rastrelliger kanagurta.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O