  • Search Results
  • Development of a new platform for secretory production of recombinant proteins in Corynebacterium glutamicum.
Development of a new platform for secretory production of recombinant proteins in Corynebacterium glutamicum.
Biotechnology and bioengineering (2015-07-03)
Sung Sun Yim, Jae Woong Choi, Roo Jin Lee, Yong Jae Lee, Se Hwa Lee, So Young Kim, Ki Jun Jeong
ABSTRACT
Corynebacterium glutamicum, which has been for long an industrial producer of various L-amino acids, nucleic acids, and vitamins, is now also regarded as a potential host for the secretory production of recombinant proteins. To harness its potential as an industrial platform for recombinant protein production, the development of an efficient secretion system is necessary. Particularly, regarding protein production in large-scale bioreactors, it would be appropriate to develop a secretory expression system that is specialized for high cell density cultivation conditions. Here we isolated a new signal peptide that mediates the efficient secretion of recombinant proteins under high cell density cultivation conditions. The secretome of C. glutamicum ATCC 13032 under high cell density cultivation conditions was initially investigated, and one major protein was identified as a hypothetical protein encoded by cg1514. Novel secretory production systems were then developed using the Cg1514 signal peptide and its own promoter. Efficient protein secretion was demonstrated using three protein models: endoxylanase, α-amylase, and camelid antibody fragment (VHH). For large-scale production, fed-batch cultivations were also conducted and high yields were successfully achieved--as high as 1.07 g/L (endoxylanase), 782.6 mg/L (α-amylase), and 1.57 g/L (VHH)--in the extracellular medium. From the culture media, all model proteins could be simply purified by one-step column chromatography with high purities and recovery yields. To the best of our knowledge, this is the first report of the development of an efficient secretory expression system by secretome analysis under high cell density cultivation conditions in C. glutamicum.
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture