  • Search Results
  • Isothermal Detection of Mycoplasma pneumoniae Directly from Respiratory Clinical Specimens.
Isothermal Detection of Mycoplasma pneumoniae Directly from Respiratory Clinical Specimens.
Journal of clinical microbiology (2015-07-17)
Brianna L Petrone, Bernard J Wolff, Alexandra A DeLaney, Maureen H Diaz, Jonas M Winchell
ABSTRACT
Mycoplasma pneumoniae is a leading cause of community-acquired pneumonia (CAP) across patient populations of all ages. We have developed a loop-mediated isothermal amplification (LAMP) assay that enables rapid, low-cost detection of M. pneumoniae from nucleic acid extracts and directly from various respiratory specimen types. The assay implements calcein to facilitate simple visual readout of positive results in approximately 1 h, making it ideal for use in primary care facilities and resource-poor settings. The analytical sensitivity of the assay was determined to be 100 fg by testing serial dilutions of target DNA ranging from 1 ng to 1 fg per reaction, and no cross-reactivity was observed against 17 other Mycoplasma species, 27 common respiratory agents, or human DNA. We demonstrated the utility of this assay by testing nucleic acid extracts (n = 252) and unextracted respiratory specimens (n = 72) collected during M. pneumoniae outbreaks and sporadic cases occurring in the United States from February 2010 to January 2014. The sensitivity of the LAMP assay was 88.5% tested on extracted nucleic acid and 82.1% evaluated on unextracted clinical specimens compared to a validated real-time PCR test. Further optimization and improvements to this method may lead to the availability of a rapid, cost-efficient laboratory test for M. pneumoniae detection that is more widely available to primary care facilities, ultimately facilitating prompt detection and appropriate responses to potential M. pneumoniae outbreaks and clusters within the community.
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, Vetec, reagent grade
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Potassium chloride, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-100.5%