  • Search Results
  • Comparing a silver-impregnated activated carbon with an unmodified activated carbon for disinfection by-product minimisation and precursor removal.
Comparing a silver-impregnated activated carbon with an unmodified activated carbon for disinfection by-product minimisation and precursor removal.
The Science of the total environment (2015-11-08)
Kalinda Watson, Maria José Farré, Nicole Knight
ABSTRACT
During disinfection, bromide, iodide and natural organic matter (NOM) in source waters can lead to the formation of brominated and/or iodinated disinfection by-products (DBPs), which are often more toxic than their chlorinated analogues. The objective of this study was to compare the efficiency of a silver-impregnated activated carbon (SIAC) with the equivalent unimpregnated granular activated carbon (GAC) for the removal of bromide, iodide and NOM from a matrix of synthetic waters with variable NOM, halide, and alkalinity concentrations, and to investigate the impact on DBP formation. An enhanced coagulation (EC) pre-treatment was employed prior to sample exposure to either carbon adsorbent. Excellent halide removals were observed by the SIAC treatment across the sample matrix, with iodide concentrations consistently reduced to below the method reporting limit (<2 μg/L) from as high as 25 μg/L, and 95±4% removal of bromide achieved. Bromide removal by unimpregnated GAC was poor, however iodide removal was comparable to that achieved by SIAC. The combination of EC with SIAC treatment removed 77±8% of the dissolved organic carbon (DOC) present, across the sample matrix, which was similar to removals by EC/GAC (67±14%). Combined EC/SIAC treatment reduced both total trihalomethanes (tTHMs) and total dihaloacetonitriles (tDHANs) formation by 97±3%, while also achieving a greater than 74% removal of two chloropropanones and a 92±8% decrease in chloral hydrate (CH), compared to untreated samples, regardless of the sample's starting water quality (bromide, alkalinity and NOM concentration). Combined EC/GAC treatment led to similar DBP removals to EC/SIAC for the fully chlorinated DBPs, however, brominated DBPs were less efficiently removed, or experienced concentration increases.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Bromodichloromethane, ≥97%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, Vetec, reagent grade, 99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)