Airway Remodeling in Preschool Children with Severe Recurrent Wheeze.
American journal of respiratory and critical care medicine (2015-05-12)
Guillaume Lezmi, Philippe Gosset, Antoine Deschildre, Rola Abou-Taam, Bruno Mahut, Nicole Beydon, Jacques de Blic
ABSTRACT
Airway wall structure in preschoolers with severe recurrent wheeze is poorly described. To describe airway wall structure and inflammation in preschoolers with severe recurrent wheeze. Flexible bronchoscopy was performed in two groups of preschoolers with severe recurrent wheeze: group 1, less than or equal to 36 months (n = 20); group 2, 36-59 months (n = 29). We assessed airway inflammation, reticular basement membrane (RBM) thickness, airway smooth muscle (ASM), mucus gland area, vascularity, and epithelial integrity. Comparisons were then made with biopsies from 21 previously described schoolchildren with severe asthma (group 3, 5-11.2 yr). RBM thickness was lower in group 1 than in group 2 (3.3 vs. 3.9 μm; P = 0.02), was correlated with age (P < 0.01; ρ = 0.62), and was higher in schoolchildren than in preschoolers (6.8 vs. 3.8 μm; P < 0.01). ASM area was lower in preschoolers than in schoolchildren (9.8% vs. 16.5%; P < 0.01). Vascularity was higher in group 1 than in group 2 (P = 0.02) and group 3 (P < 0.05). Mucus gland area was higher in preschoolers than in schoolchildren (16.4% vs. 4.6%; P < 0.01). Inflammatory cell counts in biopsies were not correlated with airway wall structure. ASM area was higher in preschoolers with atopy than without atopy (13.1% vs. 7.7%; P = 0.01). Airway morphometrics and inflammation were similar in viral and multiple-trigger wheezers. In preschoolers with severe recurrent wheeze, markers of remodeling and inflammation are unrelated, and atopy is associated with ASM. In the absence of control subjects, we cannot determine whether differences observed in RBM thickness and vascularity result from disease or normal age-related development.
Sigma-Aldrich
Solketal methacrylate, 50 wt. % in dichloromethane, contains ~280 ppm 4-tert-butylcatechol as inhibitor