  • Search Results
  • Effects of Different Demineralization-Inhibiting Methods on the Shear Bond Strength of Glass-Ceramics.
Effects of Different Demineralization-Inhibiting Methods on the Shear Bond Strength of Glass-Ceramics.
Journal of prosthodontics : official journal of the American College of Prosthodontists (2014-10-03)
Erhan Dilber, Mehmet Akın, Tevfik Yavuz, Ali Erdem
ABSTRACT
This in vitro study was undertaken to evaluate the effects of different demineralization-inhibiting methods on the shear bond strength (SBS) of glass-ceramics. Ninety extracted intact human mandibular lateral insicors were randomly divided into six equal groups. Group C was left untreated, while enamel subsurface demineralization was induced in the other groups. In group D, porcelain discs (3 mm in diameter) were cemented to demineralized enamel by using total-etch photopolymerizing luting composite resin without pretreatment. Demineralized specimens in groups F, CA, M, and I were pretreated with fluoride gel, CPP-ACP paste, microabrasion, and resin infiltration, respectively, and then porcelain discs were cemented. SBS (MPa) was calculated from the failure load (N) per bonded area (mm(2)). Fracture types were examined by optical microscopy (40× magnification). Data were analyzed with ANOVA, Tukey's test, and G-test. ANOVA revealed significant intergroup differences (p < 0.01). No significant differences in SBS (MPa) were found between groups C (19.48 ± 2.0) and I (20.02 ± 1.6). Lower SBS values were recorded in groups D (7.93 ± 0.8), F (12.51 ± 1.5), CA (17.08 ± 1.3), and M (14.84 ± 1.4). Mixed and cohesive failures were the most prevalent in groups M and I, respectively. Resin infiltration enhanced the SBS of porcelain discs bonded to demineralized enamel when compared with the other demineralization-inhibiting methods. Resin infiltration could be useful to enhance adhesion of glass-ceramics to teeth with white spot lesions.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
SAFC
HEPES
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
SAFC
HEPES
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Silicic acid, BioReagent, suitable for column chromatography, 100-200 mesh (75 - 150 μm)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Silicic acid, suitable for column chromatography, 60-200 mesh
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O