  • Search Results
  • Modulation of intestinal calcium and phosphate transport in young goats fed a nitrogen- and/or calcium-reduced diet.
Modulation of intestinal calcium and phosphate transport in young goats fed a nitrogen- and/or calcium-reduced diet.
The British journal of nutrition (2015-10-08)
Kristin Elfers, Mirja R Wilkens, Gerhard Breves, Alexandra S Muscher-Banse
ABSTRACT
Feeding ruminants a reduced N diet is a common approach to reduce N output based on rumino-hepatic circulation. However, a reduction in N intake caused massive changes in Ca and inorganic phosphate (Pi) homoeostasis in goats. Although a single dietary Ca reduction stimulated intestinal Ca absorption in a calcitriol-dependent manner, a concomitant reduction of Ca and N supply led to a decrease in calcitriol, and therefore a modulation of intestinal Ca and Pi absorption. The aim of this study was to examine the potential effects of dietary N or Ca reduction separately on intestinal Ca and Pi transport in young goats. Animals were allocated to a control, N-reduced, Ca-reduced or combined N- and Ca-reduced diet for about 6-8 weeks, whereby N content was reduced by 25 % compared with recommendations. In Ussing chamber experiments, intestinal Ca flux rates significantly decreased in goats fed a reduced N diet, whereas Pi flux rates were unaffected. In contrast, a dietary Ca reduction stimulated Ca flux rates and decreased Pi flux rates. The combined dietary N and Ca reduction withdrew the stimulating effect of dietary Ca reduction on Ca flux rates. The expression of Ca-transporting proteins decreased with a reduced N diet too, whereas Pi-transporting proteins were unaffected. In conclusion, a dietary N reduction decreased intestinal Ca transport by diminishing Ca-transporting proteins, which became clear during simultaneous N and Ca reduction. Therefore, N supply in young ruminant nutrition is of special concern for intestinal Ca transport.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Adapter for ACE Micro/Mini-Lab® kit, Distilling, Joint: ST/NS 14/10
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 1.8 gal (6.8 L)
Adapter for ACE Micro/Mini-Lab® kit, Mini adapter
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 3.6 gal (13.6 L)
Adapter for ACE Micro/Mini-Lab® kit, Vacuum-takeoff with hose barb, Joint: ST/NS 14/10
Mini chemical spill kit