  • Search Results
  • Protein-Protein Multilayer Oil-in-Water Emulsions for the Microencapsulation of Flaxseed Oil: Effect of Whey and Fish Gelatin Concentration.
Protein-Protein Multilayer Oil-in-Water Emulsions for the Microencapsulation of Flaxseed Oil: Effect of Whey and Fish Gelatin Concentration.
Journal of agricultural and food chemistry (2015-10-13)
Patrick Fustier, Allaoua Achouri, Ali R Taherian, Michel Britten, Marylène Pelletier, Hassan Sabik, Sébastien Villeneuve, Martin Mondor
ABSTRACT
The impact of whey protein isolate (WPI) and fish gelatin (FG) deposited sequentially at concentrations of 0.1, 0.5, and 0.75% on the surface of primary oil-in-water emulsions containing 5% flaxseed oil stabilized with either 0.5% fish gelatin or whey protein, respectively, was investigated. The results revealed that the adsorption of WPI/FG or FG/WPI complexes to the emulsion interface led to the formation of oil-in-water (o/w) emulsions with different stabilities and different protection degrees of the flaxseed oil. Deposition of FG on the WPI primary emulsion increased the particle size (from 0.53 to 1.58 μm) and viscosity and decreased electronegativity (from -23.91 to -11.15 mV) of the complexes. Different trends were noted with the deposition of WPI on the FG primary emulsion, resulting in decreasing particle size and increasing electronegativity and viscosity to a lower extent. Due to the superior tension-active property of WPI, the amount of protein load in the WPI primary emulsion as well as in WPI/FG complex was significantly higher than the FG counterparts. A multilayer emulsion made with 0.5% WPI/0.75% FG exhibited the lowest oxidation among all of the multilayered emulsions tested (0.32 ppm of hexanal) after 21 days, likely due to the charge effect of FG that may prevent pro-oxidant metals to interact with the flaxseed oil.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Hexanal, ≥97%, FCC, FG