  • Search Results
  • A quick method for determination of psychoactive agents in serum and hair by using capillary electrophoresis and mass spectrometry.
A quick method for determination of psychoactive agents in serum and hair by using capillary electrophoresis and mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2015-04-19)
Aneta Woźniakiewicz, Renata Wietecha-Posłuszny, Michał Woźniakiewicz, Ewelina Bryczek, Paweł Kościelniak
ABSTRACT
The aim of the research was to develop a new sensitive method for simultaneously the determination of psychoactive drugs: 1-benzylpiperazine, 7-aminoclonazepam, alprazolam, clonazepam, diazepam, estazolam, lorazepam and tetrazepam in human serum and hair samples. In the preparation step, microwave-assisted extraction (MAE) was used. Extracts were analyzed by means of capillary electrophoresis with mass spectrometry time-of-flight detection (CE-TOF-MS). In the validation study of the MAE/CE-TOF-MS analytical method, three concentration levels of analytes (10, 100 and 250 ng/mL for serum and 0.2, 2.2 and 5.6 ng/mL for hair) were taken into account. Such parameters as limit of detection (0.4-1.2 ng/mL for serum, 6.0-23.0 pg/mg for hair), limit of quantification (1.3-4.1 ng/mL for serum, 20.0-77.0 pg/mg for hair), precision (3.0-11.3% for serum, 2.4-14.2% for hair), accuracy of the assay (RE) (-8.0 to 12.0% for serum, -8.0 to 11.0% for hair), recovery (88.6-113.4% for serum, 86.1-107.4% for hair) and matrix effects (87.9-110.7% for serum, 85.1-108.4% for hair) were calculated for the studied compounds. Then, the MAE/CE-TOF-MS method was successfully applied to the analysis of hair samples taken from patients treated with benzodiazepines.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate