  • Search Results
  • Self-Assembly of Catecholic Moiety-Containing Cationic Random Acrylic Copolymers.
Self-Assembly of Catecholic Moiety-Containing Cationic Random Acrylic Copolymers.
The journal of physical chemistry. B (2015-06-16)
Vincenzo Taresco, Lorenzo Gontrani, Fernanda Crisante, Iolanda Francolini, Andrea Martinelli, Lucio D'Ilario, Federico Bordi, Antonella Piozzi
ABSTRACT
Amphiphilic polyelectrolytes (APEs), exhibiting particular self-association properties in aqueous media, can be used in different industrial applications, including drug delivery systems. Their typical core-shell structure (micelle) depends on the balance of interactions between hydrophobic and ionizable monomer units. In this work, the structure of amphiphilic cationic random copolymers, obtained by employing different molar ratios of two acrylic monomers, one bearing in the side chain a tertiary amine (N,N-diethylethylendiamine, DED) and the other one a hydrophobic catecholic group (hydroxytyrosol, HTy), was investigated by atomistic molecular dynamics (MD) simulation, (1)H NMR analysis, dynamic light scattering (DLS), and zeta potential measurements. The structures of p(AcDED-co-AcHTy) copolymers were compared with that of the cationic homopolymer (pAcDED). MD simulation showed a chain folding in water solution of all polymer materials consistent with the degree of hydrophobicity of the chain, that increases with the number of aromatic residues. This phenomenon was induced by the interaction between the charged amine groups with water and by the associated attraction between aromatic rings inside the molecule. In addition, the p(AcDED-co-AcHTy) 70/30 copolymer had a marked tendency to self-assemble as shown by the radial distribution function among catechol carbon atoms. Electrical conductivity measurements evidenced a micellar arragment for all of the synthesized copolymers, and specially for p(AcDED-co-AcHTy) 70/30, a flower micelle structure seem to be more likely. The stacking interactions among catecholic groups present in the side chain of the copolymers reduced the size and charge density specially for the p(AcDED-co-AcHTy) 70/30 copolymer. Finally, the good antimicrobial activity of all copolymers confirmed the right reached amphiphilic balance. Indeed, a considerable reduction of the minimum inhibitory concentration (from 100 μg/mL to 40 μg/mL for pAcDED and p(AcDED-co-AcHTy) 70/30, respectively) was obtained by introducing a hydrophobic group molar fraction of 0.3.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Acryloyl chloride, ≥97%, contains ~400 ppm phenothiazine as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Sodium hydrosulfite, ≥82% (RT)
Sigma-Aldrich
Acryloyl chloride, 97.0%, contains <210 ppm MEHQ as stabilizer
Sigma-Aldrich
Dimethyl carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
N,N-Diethylethylenediamine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
2-(4-Hydroxyphenyl)ethanol, 98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D