  • Search Results
  • Pentacyclic triterpene in Olea europaea L: A simultaneous determination by high-performance liquid chromatography coupled to mass spectrometry.
Pentacyclic triterpene in Olea europaea L: A simultaneous determination by high-performance liquid chromatography coupled to mass spectrometry.
Journal of chromatography. A (2015-07-27)
Estela Giménez, M Emília Juan, Sara Calvo-Melià, José Barbosa, Victoria Sanz-Nebot, Joana M Planas
ABSTRACT
Pentacyclic triterpenes are gaining interest due to their beneficial health effects, as anti-inflammatory, anti-diabetic and anti-tumoral, among others. In this study, an analytical LC-MS method was developed for the simultaneous determination of maslinic, oleanolic and ursolic acids along with erythrodiol and uvaol, which are the main triterpenic compounds present in the fruits and leaves of Olea europaea L. A Zorbax Eclipse PAH column at 30°C with mobile phase of water (17%) and methanol (83%) at 0.8mL/min conformed the optimal chromatographic conditions that allowed the separation of the compounds of interest, two pairs of which are isomers differing only in the position of one methyl group (oleanolic-ursolic and erythrodiol-uvaol). The ionization was performed in an APCI source at 450°C programmed in negative mode for the triterpenic acids, and in positive for the alcohols. An ion trap (LC-IT-MS) and a triple quadrupole (LC-QqQ-MS) were assessed for maximal sensitivity that was achieved with LC-QqQ-MS. The LODs of triterpenic acids were lower than 1nM, whereas for erythrodiol and uvaol were 4.5 and 7.5nM, respectively. The method was linear for the five analytes in the range of concentrations from 0.005 to 15μM with correlation coefficients exceeding 0.99. The precision and accuracy were ≤9.90% and ≤9.57%, respectively. The applicability of the validated method was assessed in the analysis of the pentacyclic triterpenes in Marfil table olives, after the optimization of the extraction procedure. The developed method constitutes the first step for future studies of triterpenic compounds present in foods that would allow establishing their effects on human health.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Maslinic acid, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
SAFC
Acetic acid, glacial
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate