  • Search Results
  • A polysaccharide from Andrographis paniculata induces mitochondrial-mediated apoptosis in human hepatoma cell line (HepG2).
A polysaccharide from Andrographis paniculata induces mitochondrial-mediated apoptosis in human hepatoma cell line (HepG2).
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-02-06)
Yanmei Zou, Hua Xiong, Huihua Xiong, Tao Lu, Feng Zhu, Zhiyong Luo, Xianglin Yuan, Yihua Wang
ABSTRACT
In the present study, we investigated the effects and action mechanisms of a purified polysaccharide (APWP) from Andrographis paniculata, on human hepatocellular carcinoma (HCC) HepG2 cells. The results showed that APWP was able to suppress the proliferation of HepG2 cells via inducing apoptosis. Western blot analysis revealed that dose-dependent increase in proapoptotic Bax protein and no change in antiapoptotic Bcl-2 protein in APWP-treated cells. Furthermore, exposure of tumor cells to APWP resulted in a loss of mitochondrial membrane potential (MMP) and the release of cytochrome c from the mitochondria to the cytosol. Besides, caspase-9 and caspase-3 were activated while caspase-8 was not affected in HepG2 cells followed by APWP treatment. All these results point clearly to the involvement of mitochondria-mediated signaling pathway in APWP-induced apoptosis and strongly suggest that APWP seems to be safe and effective in the prevention and treatment of HCC.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
D-(+)-Galactose, ≥99%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
D-(+)-Galactose, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-(−)-Fucose, ≥99%
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR