  • Search Results
  • Elevated concentrations of serum matrix metalloproteinase-2 and -9 and their associations with circulating markers of cardiovascular diseases in chronic arsenic-exposed individuals.
Elevated concentrations of serum matrix metalloproteinase-2 and -9 and their associations with circulating markers of cardiovascular diseases in chronic arsenic-exposed individuals.
Environmental health : a global access science source (2015-12-08)
Md Shofikul Islam, Nayan Chandra Mohanto, Md Rezaul Karim, Sharmin Aktar, Md Mominul Hoque, Atiqur Rahman, Momotaj Jahan, Rabeya Khatun, Abdul Aziz, Kazi Abdus Salam, Zahangir Alam Saud, Mostaque Hossain, Aminur Rahman, Abul Mandal, Azizul Haque, Hideki Miyataka, Seiichiro Himeno, Khaled Hossain
ABSTRACT
Cardiovascular diseases (CVDs) and cancers are the major causes of chronic arsenic exposure-related morbidity and mortality. Matrix metalloproteinase-2 (MMP-2) and -9 (MMP-9) are deeply involved in the pathogenesis of CVDs and cancers. This study has been designed to evaluate the interactions of arsenic exposure with serum MMP-2 and MMP-9 concentrations especially in relation to the circulating biomarkers of CVDs. A total of 373 human subjects, 265 from arsenic-endemic and 108 from non-endemic areas in Bangladesh were recruited for this study. Arsenic concentrations in the specimens were measured by inductively coupled plasma mass spectroscopy (ICP-MS) and serum MMPs were quantified by immunoassay kits. Serum MMP-2 and MMP-9 concentrations in arsenic-endemic population were significantly (p < 0.001) higher than those in non-endemic population. Both MMPs showed significant positive interactions with drinking water (r s  = 0.208, p < 0.001 for MMP-2; r s  = 0.163, p < 0.01 for MMP-9), hair (r s  = 0.163, p < 0.01 for MMP-2; r s  = 0.173, p < 0.01 for MMP-9) and nail (r s  = 0.160, p < 0.01 for MMP-2; r s  = 0.182, p < 0.001 for MMP-9) arsenic of the study subjects. MMP-2 concentrations were 1.02, 1.03 and 1.05 times, and MMP-9 concentrations were 1.03, 1.06 and 1.07 times greater for 1 unit increase in log-transformed water, hair and nail arsenic concentrations, respectively, after adjusting for covariates (age, sex, BMI, smoking habit and hypertension). Furthermore, both MMPs were increased dose-dependently when the study subjects were split into three (≤10, 10.1-50 and > 50 μg/L) groups based on the regulatory upper limit of water arsenic concentration set by WHO and Bangladesh Government. MMPs were also found to be significantly (p < 0.05) associated with each other. Finally, the concentrations of both MMPs were correlated with several circulating markers related to CVDs. This study showed the significant positive associations and dose-response relationships of arsenic exposure with serum MMP-2 and MMP-9 concentrations. This study also showed the interactions of MMP-2 and MMP-9 concentrations with the circulating markers of CVDs suggesting the MMP-2 and MMP-9 -mediated mechanism of arsenic-induced CVDs.
Sigma-Aldrich
Yttrium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Yttrium, wire reel, 0.05m, diameter 1.0mm, square section, 99.9%
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Arsenic, 99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Arsenic, powder, ≥99.997% trace metals basis